(2-Bromo-6-fluorophenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

116696-500MG 116696-1G 116696-5G 116696-25G
77323-580EA 161.71 USD
77323-580 77346-052 77359-702 77383-548
(2-Bromo-6-fluorophenyl)(phenyl)methanol ≥97%
(2-Bromo-6-fluorophenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD34783223
CAS Number: 1501981-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR