2-(2,3-Difluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
116688-500MG
116688-1G
116688-5G
116688-25G
77323-574EA
223.51
USD
77323-574
77347-654
77359-696
77383-542
2-(2,3-Difluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
2-(2,3-Difluorophenyl)-1,1,1-trifluoropropan-2-ol
Precaución: for Research Use Only
Formula:
C₉H₇F₅O MW: 226.1432 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34783221 CAS Number: 1512179-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...