(2,5-difluorophenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

116687-500MG 116687-1G 116687-5G
77323-572EA 348.14 USD
77323-572 77347-652 77371-688
(2,5-difluorophenyl)(phenyl)methanol ≥97%
(2,5-difluorophenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O
Storage Temperature: Ambient
CAS Number: 153877-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR