2-(2,5-Difluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%

Supplier: AOBChem USA

Warning

116686-500MG 116686-1G 116686-5G 116686-25G
77323-570EA 182.31 USD
77323-570 77347-076 77359-694 77383-540
2-(2,5-Difluorophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
2-(2,5-Difluorophenyl)-1,1,1-trifluoropropan-2-ol

Precaución: for Research Use Only

Formula: C₉H₇F₅O
MW: 226.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16067997
CAS Number: 1249339-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR