2,3-Dibromo-1-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

116681-1G 116681-5G 116681-25G
77341-450EA 86.52 USD
77341-450 77359-692 77371-686
2,3-Dibromo-1-fluoro-4-methoxybenzene ≥97%
2,3-Dibromo-1-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂FO
MW: 283.92 g/mol
Storage Temperature: Ambient
CAS Number: 1266379-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR