2-Bromo-4-iodo-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

116488-500MG 116488-1G 116488-5G 116488-25G
77323-568EA 182.31 USD
77323-568 77347-074 77359-690 77383-538
2-Bromo-4-iodo-1,3-dimethylbenzene ≥95%
2-Bromo-4-iodo-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrI
MW: 310.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738492
CAS Number: 1805106-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR