2-Bromo-4-chloro-6-fluorotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-5-chloro-3-fluoro-2-methylbenzene

116486-500MG 116486-1G 116486-5G 116486-25G
77323-566EA 241.02 USD
77323-566 77347-650 77359-688 77383-536
2-Bromo-4-chloro-6-fluorotoluene ≥97%
2-Bromo-4-chloro-6-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845453
CAS Number: 1082040-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR