5-Bromo-4-iodo-2-methoxyaniline ≥97%

Supplier: AOBChem USA

Warning

116414-500MG 116414-1G 116414-5G 116414-25G
77323-564EA 94.76 USD
77323-564 77342-516 77359-684 77371-680
5-Bromo-4-iodo-2-methoxyaniline ≥97%
5-Bromo-4-iodo-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrINO
Storage Temperature: Ambient
MDL Number: MFCD34783174
CAS Number: 2091791-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR