3-hydroxy-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

116404-500MG 116404-1G 116404-5G
77339-188EA 561.35 USD
77339-188 77347-648 77371-678
3-hydroxy-2,4-dimethoxybenzoic acid ≥97%
3-hydroxy-2,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₅
Storage Temperature: Ambient
MDL Number: MFCD20640180
CAS Number: 50389-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR