5-Fluoro-2-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Iodo-5-fluoropyridine

11614-1G 11614-5G 11614-25G
77340-940EA 64.89 USD
77340-940 77359-680 77371-676
5-Fluoro-2-iodopyridine ≥97%
5-Fluoro-2-iodopyridine

Precaución: for Research Use Only

Formula: C5H3FIN
Storage Temperature: Ambient
MDL Number: MFCD07787574
CAS Number: 159870-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR