4-(2-Fluoro-5-methylphenyl)-3,6-dihydro-2H-pyran ≥95%

Supplier: AOBChem USA

Warning

116097-500MG 116097-1G 116097-5G
77335-580EA 357.41 USD
77335-580 77347-642 77371-670
4-(2-Fluoro-5-methylphenyl)-3,6-dihydro-2H-pyran ≥95%
4-(2-Fluoro-5-methylphenyl)-3,6-dihydro-2H-pyran

Precaución: for Research Use Only

Formula: C₁₂H₁₃FO
MW: 192.2294 g/mol
Storage Temperature: Ambient
CAS Number: 2918960-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR