(4-Chloro-3-ethoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

116077-1G 116077-5G 116077-25G
77341-610EA 95.79 USD
77341-610 77359-678 77371-668
(4-Chloro-3-ethoxyphenyl)trimethylsilane ≥97%
(4-Chloro-3-ethoxyphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₁₇ClOSi
Storage Temperature: Ambient
MDL Number: MFCD34782957
CAS Number: 2901099-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR