(2-Fluoro-3-(methylthio)phenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

116064-500MG 116064-1G 116064-5G
77336-580EA 388.31 USD
77336-580 77347-640 77371-666
(2-Fluoro-3-(methylthio)phenyl)trimethylsilane ≥97%
(2-Fluoro-3-(methylthio)phenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₅FSSi
MW: 214.3750 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34782949
CAS Number: 2901109-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR