1-(2-Fluoro-3-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
115522-500MG
115522-1G
115522-5G
115522-25G
77323-556EA
202.91
USD
77323-556
77347-636
77359-676
77383-532
1-(2-Fluoro-3-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥97%
1-(2-Fluoro-3-(trifluoromethyl)phenyl)-2-methylpropan-1-ol
Precaución: for Research Use Only
Formula:
C₁₁H₁₂F₄O MW: 236.206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23823177 CAS Number: 1510678-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...