1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115506-500MG 115506-1G 115506-5G 115506-25G
77323-554EA 267.8 USD
77323-554 77347-634 77359-674 77383-530
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2-methylpropan-1-ol ≥97%
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrFO₂
MW: 277.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34782702
CAS Number: 1501657-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR