1-(2,5-Difluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

115487-500MG 115487-1G 115487-5G
77323-552EA 348.14 USD
77323-552 77347-632 77371-662
1-(2,5-Difluorophenyl)-2-methylpropan-1-ol ≥95%
1-(2,5-Difluorophenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD12787108
CAS Number: 1216069-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR