2-Methyl-1-(4-(trifluoromethyl)phenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115479-500MG 115479-1G 115479-5G 115479-25G
77323-548EA 277.07 USD
77323-548 77347-628 77359-670 77383-526
2-Methyl-1-(4-(trifluoromethyl)phenyl)propan-1-ol ≥97%
2-Methyl-1-(4-(trifluoromethyl)phenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃O
MW: 218.216 g/mol
Storage Temperature: Ambient
CAS Number: 116707-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR