1-(3-Fluorophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115474-500MG 115474-1G 115474-5G 115474-25G
77323-546EA 121.54 USD
77323-546 77343-890 77359-668 77371-660
1-(3-Fluorophenyl)-2-methylpropan-1-ol ≥97%
1-(3-Fluorophenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153466
CAS Number: 771-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR