1-(4-Fluorophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115470-500MG 115470-1G 115470-5G
77337-002EA 403.76 USD
77337-002 77347-626 77371-656
1-(4-Fluorophenyl)-2-methylpropan-1-ol ≥97%
1-(4-Fluorophenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11934573
CAS Number: 1766-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR