1-(4-Bromophenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115456-1G 115456-5G 115456-25G
77341-204EA 75.19 USD
77341-204 77359-664 77371-654
1-(4-Bromophenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(4-Bromophenyl)-2,2-dimethylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
CAS Number: 73492-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR