1-(4-Methoxyphenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115450-1G 115450-5G 115450-25G
77341-292EA 81.37 USD
77341-292 77359-662 77371-652
1-(4-Methoxyphenyl)-2-methylpropan-1-ol ≥97%
1-(4-Methoxyphenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD11934907
CAS Number: 18228-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR