4'-Chloro-2,3,4,5-tetrahydro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

115419-500MG 115419-1G
77340-450EA 1100.04 USD
77340-450 77347-624
4'-Chloro-2,3,4,5-tetrahydro-1,1'-biphenyl ≥97%
4'-Chloro-2,3,4,5-tetrahydro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₁₃Cl
MW: 192.685 g/mol
Storage Temperature: Ambient
CAS Number: 17380-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR