4-Bromo-2-iodo-6-methylphenol ≥95%

Supplier: AOBChem USA

Warning

114735-500MG 114735-1G 114735-5G 114735-25G
77323-542EA 161.71 USD
77323-542 77346-050 77359-660 77383-522
4-Bromo-2-iodo-6-methylphenol ≥95%
4-Bromo-2-iodo-6-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.93 g/mol
Storage Temperature: Ambient
CAS Number: 855157-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR