3-Chloro-4-methoxy-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11454-500MG 11454-1G 11454-5G
77338-708EA 511.91 USD
77338-708 77347-622 77371-650
3-Chloro-4-methoxy-5-methylphenylboronic acid ≥97%
3-Chloro-4-methoxy-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₃
Storage Temperature: Ambient
MDL Number: MFCD04039009
CAS Number: 1050509-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR