4-Bromo-2,6-diethylphenol ≥97%

Supplier: AOBChem USA

Warning

114416-500MG 114416-1G 114416-5G 114416-25G
77323-540EA 276.04 USD
77323-540 77347-620 77359-654 77383-520
4-Bromo-2,6-diethylphenol ≥97%
4-Bromo-2,6-diethylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
CAS Number: 63770-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR