1-Bromo-2-((4-fluorobenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

114229-1G 114229-5G 114229-25G
77341-856EA 105.06 USD
77341-856 77359-648 77371-638
1-Bromo-2-((4-fluorobenzyl)oxy)benzene ≥97%
1-Bromo-2-((4-fluorobenzyl)oxy)benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782686
CAS Number: 494772-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR