4-((4-methoxybenzyl)oxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

114009-500MG 114009-1G 114009-5G 114009-25G
77323-534EA 133.9 USD
77323-534 77344-426 77359-644 77383-516
4-((4-methoxybenzyl)oxy)benzaldehyde ≥97%
4-((4-methoxybenzyl)oxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03937574
CAS Number: 77182-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR