2,6-Dibromo-4-methylpyridin-3-amine ≥97%

Supplier: AOBChem USA

Warning

114005-500MG 114005-1G 114005-5G 114005-25G
77323-532EA 117.42 USD
77323-532 77343-518 77359-642 77371-636
2,6-Dibromo-4-methylpyridin-3-amine ≥97%
2,6-Dibromo-4-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆Br₂N₂
MW: 265.933 g/mol
Storage Temperature: Ambient
CAS Number: 126354-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR