8-(4-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

113998-1G 113998-5G 113998-25G
77341-814EA 104.03 USD
77341-814 77359-640 77371-634
8-(4-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(4-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₇BrO₃
MW: 313.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410871
CAS Number: 125962-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR