2,3-Dibromo-4,6-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

113744-500MG 113744-1G 113744-5G
77323-530EA 344.02 USD
77323-530 77347-616 77371-632
2,3-Dibromo-4,6-dimethylphenol ≥97%
2,3-Dibromo-4,6-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.957 g/mol
Storage Temperature: Ambient
CAS Number: 83755-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR