2,3-Dibromo-6-fluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

113724-500MG 113724-1G 113724-5G
77323-526EA 347.11 USD
77323-526 77347-614 77371-630
2,3-Dibromo-6-fluorobenzonitrile ≥97%
2,3-Dibromo-6-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂Br₂FN
MW: 278.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784673
CAS Number: 1805122-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR