1,3-Dibromo-5-(difluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

113686-500MG 113686-1G 113686-5G 113686-25G
77323-524EA 82.4 USD
77323-524 77342-172 77359-634 77371-626
1,3-Dibromo-5-(difluoromethyl)benzene ≥97%
1,3-Dibromo-5-(difluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₄Br₂F₂
MW: 285.911 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698564
CAS Number: 1214388-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR