2-(Cyclopent-1-en-1-yl)-4-fluoro-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

113517-500MG 113517-1G 113517-5G 113517-25G
77323-520EA 249.26 USD
77323-520 77347-610 77359-630 77383-510
2-(Cyclopent-1-en-1-yl)-4-fluoro-1-methoxybenzene ≥95%
2-(Cyclopent-1-en-1-yl)-4-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₃FO
MW: 192.229 g/mol
Storage Temperature: Ambient
CAS Number: 1233530-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR