1-(Cyclopent-1-en-1-yl)-4-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

113510-500MG 113510-1G 113510-5G
77336-296EA 384.19 USD
77336-296 77347-608 77371-622
1-(Cyclopent-1-en-1-yl)-4-ethylbenzene ≥95%
1-(Cyclopent-1-en-1-yl)-4-ethylbenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₆
MW: 172.266 g/mol
Storage Temperature: Ambient
CAS Number: 19099-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR