1-(Cyclopent-1-en-1-yl)-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

113506-500MG 113506-1G 113506-5G
77338-070EA 461.44 USD
77338-070 77347-606 77371-620
1-(Cyclopent-1-en-1-yl)-2-methoxybenzene ≥97%
1-(Cyclopent-1-en-1-yl)-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₄O
MW: 174.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01035656
CAS Number: 39877-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR