1-(Cyclopent-1-en-1-yl)-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

113505-500MG 113505-1G
77340-566EA 1971.42 USD
77340-566 77347-604
1-(Cyclopent-1-en-1-yl)-2-fluorobenzene ≥97%
1-(Cyclopent-1-en-1-yl)-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₁F
Storage Temperature: Ambient
MDL Number: MFCD15527431
CAS Number: 166878-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR