1-(Cyclopent-1-en-1-yl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

113503-500MG 113503-1G 113503-5G
77323-518EA 326.51 USD
77323-518 77347-602 77371-618
1-(Cyclopent-1-en-1-yl)-4-methylbenzene ≥97%
1-(Cyclopent-1-en-1-yl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₄
MW: 158.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527426
CAS Number: 827-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR