1-(Cyclopent-1-en-1-yl)-4-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

113501-500MG 113501-1G 113501-5G
77340-146EA 792.07 USD
77340-146 77347-600 77371-616
1-(Cyclopent-1-en-1-yl)-4-ethoxybenzene ≥97%
1-(Cyclopent-1-en-1-yl)-4-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₆O
MW: 188.266 g/mol
Storage Temperature: Ambient
CAS Number: 60940-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR