4-(Cyclopent-1-en-1-yl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

113499-500MG 113499-1G 113499-5G
77335-920EA 367.71 USD
77335-920 77347-598 77371-614
4-(Cyclopent-1-en-1-yl)-1,1'-biphenyl ≥97%
4-(Cyclopent-1-en-1-yl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₇H₁₆
MW: 220.309 g/mol
Storage Temperature: Ambient
CAS Number: 415898-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR