2-(Cyclopent-1-en-1-yl)-1,4-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

113495-500MG 113495-1G 113495-5G
77337-804EA 444.96 USD
77337-804 77347-596 77371-612
2-(Cyclopent-1-en-1-yl)-1,4-dimethylbenzene ≥95%
2-(Cyclopent-1-en-1-yl)-1,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₆
MW: 172.266 g/mol
Storage Temperature: Ambient
CAS Number: 2636094-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR