1-(Cyclopent-1-en-1-yl)-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

113476-500MG 113476-1G
77340-504EA 1331.79 USD
77340-504 77347-592
1-(Cyclopent-1-en-1-yl)-4-methoxybenzene ≥97%
1-(Cyclopent-1-en-1-yl)-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₄O
MW: 174.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00456851
CAS Number: 709-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR