4-(Cyclopent-1-en-1-yl)-1-fluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

113472-500MG 113472-1G 113472-5G
77337-558EA 421.27 USD
77337-558 77347-590 77371-610
4-(Cyclopent-1-en-1-yl)-1-fluoro-2-methylbenzene ≥95%
4-(Cyclopent-1-en-1-yl)-1-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₃F
MW: 176.23 g/mol
Storage Temperature: Ambient
CAS Number: 2901082-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR