3-Ethylquinoline ≥97%

Supplier: AOBChem USA

Warning

113444-500MG 113444-1G 113444-5G 113444-25G
77323-516EA 265.74 USD
77323-516 77347-588 77359-626 77383-508
3-Ethylquinoline ≥97%
3-Ethylquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD00190587
CAS Number: 1873-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR