5-(thiophen-2-yl)pyrimidin-2-amine ≥97%

Supplier: AOBChem USA

Warning

113440-500MG 113440-1G 113440-25G 113440-5G
77323-512EA 302.82 USD
77323-512 77347-584 77383-504 77371-604
5-(thiophen-2-yl)pyrimidin-2-amine ≥97%
5-(thiophen-2-yl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₈H₇N₃S
MW: 177.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11877275
CAS Number: 198078-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR