5,6-Dichloro-N-cyclopropylnicotinamide ≥95%

Supplier: AOBChem USA

Warning

113404-500MG 113404-1G 113404-5G 113404-25G
77323-510EA 191.58 USD
77323-510 77347-582 77359-624 77383-502
5,6-Dichloro-N-cyclopropylnicotinamide ≥95%
5,6-Dichloro-N-cyclopropylnicotinamide

Precaución: for Research Use Only

Formula: C₉H₈Cl₂N₂O
MW: 231.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04695184
CAS Number: 25764-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR