4-(tert-Butyl)-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

112839-500MG 112839-1G 112839-5G
77323-502EA 322.39 USD
77323-502 77347-568 77371-596
4-(tert-Butyl)-2-isopropoxybenzoic acid ≥97%
4-(tert-Butyl)-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
MW: 236.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28357050
CAS Number: 1369829-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR