3-(Ethylthio)-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

112485-500MG 112485-1G 112485-5G 112485-25G
77323-498EA 143.17 USD
77323-498 77344-996 77359-612 77383-492
3-(Ethylthio)-2,4-difluorobenzaldehyde ≥97%
3-(Ethylthio)-2,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂OS
MW: 202.221 g/mol
Storage Temperature: Ambient
CAS Number: 2918967-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR