4-(Ethylthio)-2,3-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

112444-1G 112444-5G 112444-25G
77341-854EA 105.06 USD
77341-854 77359-608 77371-586
4-(Ethylthio)-2,3-difluorobenzaldehyde ≥97%
4-(Ethylthio)-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂OS
MW: 202.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34781549
CAS Number: 1975179-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR