1-Bromo-3-chloro-4-iodo-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

112412-1G 112412-5G 112412-25G
77341-710EA 100.94 USD
77341-710 77359-606 77371-584
1-Bromo-3-chloro-4-iodo-2-methylbenzene ≥95%
1-Bromo-3-chloro-4-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClI
MW: 331.376 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30478263
CAS Number: 1799612-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR