2-Fluoro-4-iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11218-1G 11218-5G 11218-25G
77340-868EA 61.8 USD
77340-868 77359-602 77371-578
2-Fluoro-4-iodophenylboronic acid ≥97%
2-Fluoro-4-iodophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BFIO₂
Storage Temperature: Ambient
MDL Number: MFCD02093073
CAS Number: 374790-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR